Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35B1
Entrez Gene ID
10237
Gene Name
solute carrier family 35 member B1
Gene Aliases
UGTREL1
Chromosome Location
Chr.17: 49700934 - 49708173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278784.1 NP_001265713.1 6 - 7 749 60
NM_005827.2 NP_005818.2 6 - 7 774 60
XM_005256929.3 XP_005256986.1 7 - 8 689 60
XM_006721632.3 XP_006721695.1 7 - 8 893 60
XM_011524179.2 XP_011522481.1 5 - 6 659 60
XM_011524180.2 XP_011522482.1 6 - 7 625 60
GenBank mRNA AK098056.1 - 4 - 5 1849 60
BC011888.2 - 6 - 7 753 60
BM476321.1 - 2 - 3 184 60
BX641166.1 - 6 - 7 1028 60
CR745911.1 - 4 - 5 358 60
CX164572.1 - 4 - 5 381 60
D87989.1 - 6 - 7 743 60

View Full Product Details