Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35B1
Entrez Gene ID
10237
Gene Name
solute carrier family 35 member B1
Gene Aliases
UGTREL1
Chromosome Location
Chr.17: 49700934 - 49708173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278784.1 NP_001265713.1 1 - 2 219 78
NM_005827.2 NP_005818.2 1 - 2 219 78
GenBank mRNA AK293563.1 - 1 - 2 219 78
AK296073.1 - 1 - 2 188 78
AU099388.1 - 1 - 2 223 78
AU309730.1 - 1 - 2 253 78
BC011888.2 - 1 - 2 198 78
D87989.1 - 1 - 2 188 78

View Full Product Details