The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267041.2 | NP_001253970.1 | 12 - 13 | 1428 | 66 |
NM_001267042.2 | NP_001253971.1 | 12 - 13 | 1320 | 66 | |
NM_001282342.1 | NP_001269271.1 | 11 - 12 | 1351 | 66 | |
NM_015396.5 | NP_056211.2 | 14 - 15 | 1597 | 66 | |
XM_006713564.3 | XP_006713627.1 | 13 - 14 | 1516 | 66 | |
XM_006713565.2 | XP_006713628.1 | 14 - 15 | 1557 | 66 | |
XM_006713566.3 | XP_006713629.1 | 12 - 13 | 1411 | 66 | |
XM_006713567.3 | XP_006713630.1 | 13 - 14 | 1516 | 66 | |
XM_011512632.1 | XP_011510934.1 | 13 - 14 | 1464 | 66 | |
XM_011512633.1 | XP_011510935.1 | 13 - 14 | 1518 | 66 | |
XM_017006095.1 | XP_016861584.1 | 13 - 14 | 1318 | 66 | |
XM_017006096.1 | XP_016861585.1 | 12 - 13 | 1411 | 66 | |
XM_017006097.1 | XP_016861586.1 | 12 - 13 | 1264 | 66 | |
XM_017006098.1 | XP_016861587.1 | 11 - 12 | 1210 | 66 | |
XM_017006099.1 | XP_016861588.1 | 12 - 13 | 1301 | 66 | |
XM_017006100.1 | XP_016861589.1 | 13 - 14 | 1516 | 66 | |
GenBank mRNA | AK001721.1 | - | 12 - 13 | 1159 | 66 |
AK296757.1 | - | 12 - 13 | 1051 | 66 | |
AK296837.1 | - | 12 - 13 | 1128 | 66 | |
AK299229.1 | - | 8 - 9 | 789 | 66 | |
AK299769.1 | - | 11 - 12 | 1071 | 66 | |
AK304558.1 | - | 12 - 13 | 1104 | 66 | |
AK315932.1 | - | 8 - 9 | 794 | 66 | |
BC032661.1 | - | 14 - 15 | 1461 | 66 | |
BC041699.1 | - | 14 - 15 | 1276 | 66 |