Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
Chromosome Location
Chr.7: 38383694 - 38631567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001635.3 NP_001626.1 12 - 13 1351 83
NM_139316.2 NP_647477.1 12 - 13 1351 83
GenBank mRNA AF034996.1 - 12 - 13 1204 83
AF498096.1 - 3 - 4 405 83
AK291447.1 - 12 - 13 1155 83
BC034376.1 - 12 - 13 1142 83
U07616.1 - 12 - 13 1245 83
X81438.1 - 12 - 13 1209 83

View Full Product Details