Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
Chromosome Location
Chr.7: 38383694 - 38631567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139316.2 NP_647477.1 16 - 17 1488 71
XM_006715689.3 XP_006715752.1 16 - 17 3151 71
XM_006715690.3 XP_006715753.1 16 - 17 1933 71
XM_017011996.1 XP_016867485.1 16 - 17 2239 71
GenBank mRNA AF034996.1 - 16 - 17 1341 71
AF498092.1 - 5 - 6 669 71
AF498093.1 - 3 - 4 626 71
AF498094.1 - 3 - 4 1538 71
AF498096.1 - 8 - 9 1064 71

View Full Product Details