Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
Chromosome Location
Chr.7: 38383694 - 38631567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001635.3 NP_001626.1 18 - 19 1822 60
NM_139316.2 NP_647477.1 17 - 18 1696 60
XM_006715689.3 XP_006715752.1 17 - 18 3359 60
XM_006715690.3 XP_006715753.1 17 - 18 2141 60
XM_011515271.2 XP_011513573.1 18 - 19 2267 60
XM_017011995.1 XP_016867484.1 18 - 19 3485 60
XM_017011996.1 XP_016867485.1 17 - 18 2447 60
GenBank mRNA AF034996.1 - 17 - 18 1549 60
AF498092.1 - 6 - 7 877 60
AF498093.1 - 4 - 5 834 60
AF498094.1 - 4 - 5 1746 60
AF498095.1 - 5 - 6 633 60
AF498096.1 - 9 - 10 1272 60
AK291447.1 - 18 - 19 1626 60
BC034376.1 - 18 - 19 1613 60
U07616.1 - 18 - 19 1716 60
X81438.1 - 18 - 19 1680 60

View Full Product Details