The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001118887.1 | NP_001112359.1 | 1 - 2 | 617 | 103 |
NM_001147.2 | NP_001138.1 | 1 - 2 | 617 | 103 | |
GenBank mRNA | AB009865.1 | - | 1 - 2 | 316 | 103 |
AF004327.1 | - | 1 - 2 | 637 | 103 | |
AF218015.1 | - | 1 - 2 | 370 | 103 | |
AK075219.1 | - | 1 - 2 | 599 | 103 | |
AK225698.1 | - | 1 - 2 | 599 | 103 | |
AK290070.1 | - | 1 - 2 | 599 | 103 | |
AK310171.1 | - | 1 - 2 | 599 | 103 | |
AK312541.1 | - | 1 - 2 | 599 | 103 | |
BC126200.1 | - | 1 - 2 | 345 | 103 | |
BC126202.1 | - | 1 - 2 | 345 | 103 | |
BC143902.1 | - | 1 - 2 | 345 | 103 |