The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016201.3 | NP_057285.3 | 8 - 9 | 2413 | 85 |
XM_017006580.1 | XP_016862069.1 | 8 - 9 | 2270 | 85 | |
XM_017006581.1 | XP_016862070.1 | 8 - 9 | 2451 | 85 | |
XM_017006582.1 | XP_016862071.1 | 8 - 9 | 2278 | 85 | |
GenBank mRNA | AB023206.1 | - | 8 - 9 | 2346 | 85 |
AK074731.1 | - | 3 - 4 | 811 | 85 | |
AK091040.1 | - | 5 - 6 | 1481 | 85 | |
AK291616.1 | - | 8 - 9 | 2357 | 85 |