The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080510.4 | NP_001073979.3 | 1 - 2 | 344 | 111 |
XM_006721675.1 | XP_006721738.1 | 1 - 2 | 288 | 111 | |
GenBank mRNA | AL522160.3 | - | 1 - 2 | 329 | 111 |
BC045819.1 | - | 1 - 2 | 306 | 111 | |
BX403283.2 | - | 1 - 2 | 155 | 111 |