The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017392.4 | NP_001017392.2 | 6 - 7 | 2596 | 128 |
NM_001321697.1 | NP_001308626.1 | 6 - 7 | 2752 | 128 | |
NM_001321698.1 | NP_001308627.1 | 6 - 7 | 2544 | 128 | |
NM_001321699.1 | NP_001308628.1 | 6 - 7 | 2544 | 128 | |
NM_014884.4 | NP_055699.2 | 6 - 7 | 2596 | 128 | |
XM_017026137.1 | XP_016881626.1 | 6 - 7 | 2752 | 128 | |
GenBank mRNA | AB002363.2 | - | 6 - 7 | 2748 | 128 |
AF518874.1 | - | 6 - 7 | 2585 | 128 | |
AK291275.1 | - | 6 - 7 | 2563 | 128 | |
AK299860.1 | - | 6 - 7 | 2011 | 128 | |
AL832488.1 | - | 6 - 7 | 2516 | 128 | |
BC020586.1 | - | 6 - 7 | 2516 | 128 | |
BX647813.1 | - | 6 - 7 | 2525 | 128 | |
BX647933.1 | - | 2 - 3 | 233 | 129 |