Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
Chromosome Location
Chr.18: 63327736 - 63367273 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002035.2 NP_002026.1 1 - 2 261 83
XM_005266677.2 XP_005266734.1 1 - 2 228 83
GenBank mRNA AK025120.1 - 1 - 2 194 83
AK297670.1 - 1 - 2 192 83
AK312360.1 - 1 - 2 189 83
BC008797.2 - 1 - 2 240 83
BI458411.1 - 1 - 2 270 83
BT006782.1 - 1 - 2 106 83
X63657.1 - 1 - 2 213 83

View Full Product Details