Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
Chromosome Location
Chr.16: 77282128 - 77435114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326358.1 NP_001313287.1 21 - 22 3828 93
NM_199355.3 NP_955387.1 21 - 22 3824 93
XM_011522923.1 XP_011521225.1 19 - 20 3235 93
XM_017022988.1 XP_016878477.1 18 - 19 3071 93
XM_017022989.1 XP_016878478.1 18 - 19 2868 93
GenBank mRNA AK122677.1 - 6 - 7 1125 93
AK131403.1 - 18 - 19 2780 93
AY172951.1 - 6 - 7 903 93
BC063283.1 - 21 - 22 3824 93

View Full Product Details