Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
Chromosome Location
Chr.16: 74873568 - 74985146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324171.1 NP_001311100.1 8 - 9 777 63
NM_001324172.1 NP_001311101.1 8 - 9 777 63
NM_030581.3 NP_085058.3 8 - 9 750 63
XM_005256146.3 XP_005256203.1 8 - 9 754 63
XM_011523331.2 XP_011521633.1 7 - 8 594 63
XM_011523332.2 XP_011521634.1 2 - 3 177 63
XM_017023667.1 XP_016879156.1 7 - 8 611 63
XM_017023668.1 XP_016879157.1 6 - 7 387 63
XM_017023669.1 XP_016879158.1 7 - 8 482 63
XM_017023670.1 XP_016879159.1 6 - 7 490 63
GenBank mRNA AB067510.1 - 3 - 4 223 63
AK302557.1 - 8 - 9 818 63
BC004519.2 - 8 - 9 714 63
BC014887.2 - 2 - 3 142 63
BU850134.1 - 8 - 9 64

View Full Product Details