Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
Chromosome Location
Chr.16: 74873568 - 74985146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324171.1 NP_001311100.1 4 - 5 452 68
NM_001324172.1 NP_001311101.1 4 - 5 452 68
NM_030581.3 NP_085058.3 4 - 5 425 68
XM_005256146.3 XP_005256203.1 4 - 5 429 68
XM_011523331.2 XP_011521633.1 3 - 4 269 68
XM_017023667.1 XP_016879156.1 3 - 4 269 68
XM_017023668.1 XP_016879157.1 3 - 4 151 68
XM_017023669.1 XP_016879158.1 3 - 4 157 68
XM_017023670.1 XP_016879159.1 2 - 3 165 68
GenBank mRNA AK302557.1 - 4 - 5 493 68
BC004519.2 - 4 - 5 389 68
BU850134.1 - 4 - 5 377 68

View Full Product Details