Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARHGAP33
- Entrez Gene ID
-
115703
- Gene Name
- Rho GTPase activating protein 33
- Gene Aliases
-
NOMA-GAP, SNX26, TCGAP
- Chromosome Location
-
Chr.19: 35775515 - 35788822 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY044864.1 |
- |
5 - 6 |
599 |
112 |
View Full Product Details