The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322123.1 | NP_001309052.1 | 13 - 14 | 1341 | 109 |
NM_001322124.1 | NP_001309053.1 | 13 - 14 | 1210 | 109 | |
NM_001322127.1 | NP_001309056.1 | 12 - 13 | 1336 | 109 | |
NM_144709.3 | NP_653310.2 | 13 - 14 | 1398 | 109 | |
XM_011532568.2 | XP_011530870.1 | 14 - 15 | 1469 | 109 | |
XM_011532570.1 | XP_011530872.1 | 13 - 14 | 1176 | 109 | |
XM_011532571.1 | XP_011530873.1 | 13 - 14 | 1350 | 109 | |
XM_011532572.1 | XP_011530874.1 | 13 - 14 | 1270 | 109 | |
XM_011532573.2 | XP_011530875.1 | 13 - 14 | 1230 | 109 | |
XM_011532574.2 | XP_011530876.1 | 13 - 14 | 1291 | 109 | |
XM_011532576.2 | XP_011530878.1 | 13 - 14 | 1350 | 109 | |
XM_011532578.1 | XP_011530880.1 | 13 - 14 | 1328 | 109 | |
XM_017003428.1 | XP_016858917.1 | 12 - 13 | 1167 | 109 | |
XM_017003429.1 | XP_016858918.1 | 12 - 13 | 963 | 109 | |
GenBank mRNA | AK056874.1 | - | 13 - 14 | 1212 | 109 |
AK291729.1 | - | 13 - 14 | 1186 | 109 | |
AL832208.1 | - | 11 - 12 | 3737 | 109 | |
BC101680.1 | - | 13 - 14 | 1190 | 109 | |
BC101706.1 | - | 13 - 14 | 1190 | 109 | |
BC143480.1 | - | 12 - 13 | 1128 | 109 |