Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 12 - 13 2227 101
NM_001005273.2 NP_001005273.1 12 - 13 2198 101
NM_005852.3 NP_005843.2 12 - 13 2198 101
XM_005256427.4 XP_005256484.1 12 - 13 2268 101
XM_005256428.4 XP_005256485.1 12 - 13 2268 101
XM_005256429.4 XP_005256486.1 12 - 13 2268 101
XM_005256431.4 XP_005256488.1 12 - 13 2268 101
XM_006721423.3 XP_006721486.1 12 - 13 2268 101
XM_006721424.3 XP_006721487.1 12 - 13 2243 101
XM_006721428.3 XP_006721491.1 12 - 13 2268 101
XM_017024061.1 XP_016879550.1 12 - 13 2250 101
XM_017024062.1 XP_016879551.1 12 - 13 2225 101
XM_017024063.1 XP_016879552.1 12 - 13 2183 101
XM_017024064.1 XP_016879553.1 12 - 13 2243 101
XM_017024065.1 XP_016879554.1 11 - 12 2064 101
XM_017024066.1 XP_016879555.1 13 - 14 2003 101
XM_017024067.1 XP_016879556.1 13 - 14 2186 101
XM_017024068.1 XP_016879557.1 12 - 13 2159 101
XM_017024069.1 XP_016879558.1 12 - 13 2050 101
XM_017024070.1 XP_016879559.1 12 - 13 2225 101
GenBank mRNA U91543.1 - 12 - 13 2198 101

View Full Product Details