Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 26 - 27 4312 62
NM_001005273.2 NP_001005273.1 26 - 27 4283 62
NM_005852.3 NP_005843.2 26 - 27 4283 62
XM_005256427.4 XP_005256484.1 26 - 27 4353 62
XM_005256428.4 XP_005256485.1 26 - 27 4353 62
XM_005256429.4 XP_005256486.1 26 - 27 4353 62
XM_006721423.3 XP_006721486.1 26 - 27 4353 62
XM_006721424.3 XP_006721487.1 26 - 27 4328 62
XM_006721428.3 XP_006721491.1 26 - 27 4353 62
XM_017024061.1 XP_016879550.1 26 - 27 4335 62
XM_017024062.1 XP_016879551.1 26 - 27 4310 62
XM_017024063.1 XP_016879552.1 26 - 27 4268 62
XM_017024064.1 XP_016879553.1 26 - 27 4328 62
XM_017024065.1 XP_016879554.1 25 - 26 4149 62
XM_017024066.1 XP_016879555.1 27 - 28 4088 62
XM_017024067.1 XP_016879556.1 27 - 28 4271 62
XM_017024068.1 XP_016879557.1 26 - 27 4244 62
XM_017024069.1 XP_016879558.1 26 - 27 4135 62
XM_017024070.1 XP_016879559.1 26 - 27 4310 62
GenBank mRNA AK125928.1 - 3 - 4 696 62
AK296937.1 - 8 - 9 1077 62
AK308362.1 - 4 - 5 443 62
BC110648.1 - 2 - 3 116 62
U91543.1 - 26 - 27 4283 62

View Full Product Details