Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 27 - 28 4406 91
NM_001005273.2 NP_001005273.1 27 - 28 4377 91
NM_005852.3 NP_005843.2 27 - 28 4377 91
XM_005256427.4 XP_005256484.1 27 - 28 4447 91
XM_005256428.4 XP_005256485.1 27 - 28 4447 91
XM_005256429.4 XP_005256486.1 27 - 28 4447 91
XM_006721423.3 XP_006721486.1 27 - 28 4447 91
XM_006721424.3 XP_006721487.1 27 - 28 4422 91
XM_006721428.3 XP_006721491.1 27 - 28 4447 91
XM_017024061.1 XP_016879550.1 27 - 28 4429 91
XM_017024062.1 XP_016879551.1 27 - 28 4404 91
XM_017024063.1 XP_016879552.1 27 - 28 4362 91
XM_017024064.1 XP_016879553.1 27 - 28 4422 91
XM_017024065.1 XP_016879554.1 26 - 27 4243 91
XM_017024066.1 XP_016879555.1 28 - 29 4182 91
XM_017024067.1 XP_016879556.1 28 - 29 4365 91
XM_017024068.1 XP_016879557.1 27 - 28 4338 91
XM_017024069.1 XP_016879558.1 27 - 28 4229 91
XM_017024070.1 XP_016879559.1 27 - 28 4404 91
GenBank mRNA AK125928.1 - 4 - 5 790 91
AK296937.1 - 9 - 10 1171 91
AK308362.1 - 5 - 6 537 91
BC110648.1 - 3 - 4 210 91
U91543.1 - 27 - 28 4377 91

View Full Product Details