The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005587.3 | NP_005578.2 | 5 - 6 | 799 | 150 |
XM_011521581.2 | XP_011519883.1 | 5 - 6 | 1027 | 150 | |
XM_011521582.2 | XP_011519884.1 | 6 - 7 | 1232 | 150 | |
XM_017022190.1 | XP_016877679.1 | 4 - 5 | 653 | 150 | |
XM_017022191.1 | XP_016877680.1 | 4 - 5 | 645 | 150 | |
XM_017022193.1 | XP_016877682.1 | 5 - 6 | 837 | 150 | |
XM_017022194.1 | XP_016877683.1 | 4 - 5 | 653 | 150 | |
XM_017022195.1 | XP_016877684.1 | 4 - 5 | 510 | 150 | |
XM_017022197.1 | XP_016877686.1 | 5 - 6 | 2944 | 150 | |
XM_017022198.1 | XP_016877687.1 | 6 - 7 | 1153 | 150 | |
XM_017022200.1 | XP_016877689.1 | 5 - 6 | 1224 | 150 | |
XM_017022201.1 | XP_016877690.1 | 4 - 5 | 847 | 150 | |
XM_017022203.1 | XP_016877692.1 | 5 - 6 | 1027 | 150 | |
GenBank mRNA | BC013437.2 | - | 5 - 6 | 717 | 150 |
KU178181.1 | - | 3 - 4 | 398 | 150 | |
KU178182.1 | - | 3 - 4 | 398 | 150 | |
X63381.1 | - | 4 - 5 | 626 | 150 | |
X68505.1 | - | 5 - 6 | 812 | 150 |