Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C18orf15
- Entrez Gene ID
-
147276
- Gene Name
- chromosome 18 open reading frame 15
- Gene Aliases
-
HsT3231
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK055900.1 |
- |
1 - 1 |
2441 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB321566.1 |
- |
1 |
|
|
Chr. 18 - 13241591 - 13242076 [-] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
centrosomal protein 192 |
CEP192 |
PPP1R62 |
-114.5kb
|
Sense |
Chr. - 12991307 - 13125053 on Build GRCh38 |
Downstream |
LDLRAD4 antisense RNA 1 |
LDLRAD4-AS1 |
|
177.4kb
|
Anti-sense |
Chr. - 13419499 - 13427534 on Build GRCh38 |
View Full Product Details