Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 40439721 - 40468242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014952.4 NP_055767.3 2 - 3 1569 72
XM_011521366.2 XP_011519668.2 2 - 3 1693 72
XM_011521368.2 XP_011519670.2 2 - 3 1692 72
XM_011521369.2 XP_011519671.1 2 - 3 1547 72
GenBank mRNA AL833923.1 - 1 - 2 661 72
BC022782.2 - 2 - 3 1508 72

View Full Product Details