The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252335.1 | NP_001239264.1 | 7 - 8 | 2248 | 99 |
NM_032866.4 | NP_116255.2 | 6 - 7 | 2133 | 99 | |
XM_005254726.3 | XP_005254783.1 | 6 - 7 | 2151 | 99 | |
XM_005254727.4 | XP_005254784.1 | 7 - 8 | 2529 | 99 | |
XM_011522120.1 | XP_011520422.1 | 7 - 8 | 2187 | 99 | |
XM_011522121.1 | XP_011520423.1 | 6 - 7 | 2310 | 99 | |
XM_017022685.1 | XP_016878174.1 | 6 - 7 | 2768 | 99 | |
XM_017022686.1 | XP_016878175.1 | 6 - 7 | 2766 | 99 | |
GenBank mRNA | AB051536.2 | - | 5 - 6 | 1292 | 99 |
AK056673.1 | - | 6 - 7 | 2133 | 99 | |
AK307410.1 | - | 6 - 7 | 2133 | 99 | |
AY274808.1 | - | 6 - 7 | 2131 | 99 | |
BC093827.1 | - | 5 - 6 | 2075 | 99 | |
BC112049.1 | - | 5 - 6 | 2087 | 99 | |
BC118918.1 | - | 5 - 6 | 2074 | 99 |