The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286216.1 | NP_001273145.1 | 5 - 6 | 588 | 63 |
NM_019096.4 | NP_061969.3 | 5 - 6 | 783 | 63 | |
XM_017010976.1 | XP_016866465.1 | 5 - 6 | 777 | 63 | |
GenBank mRNA | AF168990.1 | - | 5 - 6 | 580 | 63 |
AK290267.1 | - | 5 - 6 | 588 | 63 | |
AL834331.1 | - | 5 - 6 | 565 | 63 | |
BC020980.2 | - | 1 - 2 | 199 | 63 | |
BC028347.2 | - | 3 - 4 | 491 | 63 | |
BC064968.1 | - | 5 - 6 | 736 | 63 |