The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193329.1 | NP_001180258.1 | 13 - 14 | 2366 | 82 |
NM_032823.5 | NP_116212.3 | 12 - 13 | 2152 | 82 | |
XM_005252284.2 | XP_005252341.1 | 12 - 13 | 2217 | 82 | |
XM_006717306.1 | XP_006717369.1 | 14 - 15 | 2514 | 82 | |
XM_011519119.1 | XP_011517421.1 | 14 - 15 | 2514 | 82 | |
XM_011519121.2 | XP_011517423.1 | 14 - 15 | 2514 | 82 | |
XM_011519132.1 | XP_011517434.1 | 11 - 12 | 1240 | 82 | |
XM_017015229.1 | XP_016870718.1 | 14 - 15 | 2557 | 82 | |
XM_017015230.1 | XP_016870719.1 | 14 - 15 | 2514 | 82 | |
XM_017015231.1 | XP_016870720.1 | 12 - 13 | 2217 | 82 | |
XM_017015233.1 | XP_016870722.1 | 14 - 15 | 2544 | 82 | |
XM_017015234.1 | XP_016870723.1 | 13 - 14 | 2347 | 82 | |
XM_017015235.1 | XP_016870724.1 | 13 - 14 | 2347 | 82 | |
XM_017015236.1 | XP_016870725.1 | 11 - 12 | 2050 | 82 | |
XM_017015237.1 | XP_016870726.1 | 11 - 12 | 2050 | 82 | |
GenBank mRNA | AJ560639.1 | - | 13 - 14 | 2231 | 82 |
AK027581.1 | - | 11 - 12 | 1403 | 82 | |
AK298959.1 | - | 4 - 5 | 409 | 82 | |
AK300486.1 | - | 3 - 4 | 254 | 82 | |
BC020194.1 | - | 13 - 14 | 1577 | 82 | |
BC029616.1 | - | 12 - 13 | 2109 | 82 | |
BP274281.1 | - | 4 - 5 | 326 | 82 | |
BX648970.1 | - | 12 - 13 | 1452 | 82 |