Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSPAN3
Entrez Gene ID
10099
Gene Name
tetraspanin 3
Gene Aliases
TM4-A, TM4SF8, TSPAN-3
Chromosome Location
Chr.15: 77044017 - 77071228 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168412.1 NP_001161884.1 4 - 5 665 117
NM_005724.5 NP_005715.1 5 - 6 857 117
NM_198902.2 NP_944492.1 4 - 5 782 117
XM_017021857.1 XP_016877346.1 5 - 6 466 117
GenBank mRNA AF054840.1 - 5 - 6 583 117
AF133423.1 - 5 - 6 657 117
AK001305.1 - 5 - 6 703 117
AK001310.1 - 5 - 6 689 117
AK001326.1 - 5 - 6 800 117
AK027793.1 - 2 - 3 1908 117
AK075336.1 - 5 - 6 738 117
AK075482.1 - 4 - 5 663 117
AK293677.1 - 6 - 7 646 117
AK298157.1 - 4 - 5 546 117
BC000704.2 - 5 - 6 758 117
BC004280.1 - 5 - 6 719 117
BC009248.2 - 5 - 6 722 117
BC011206.1 - 5 - 6 751 117
M69023.1 - 1 - 2 132 117

View Full Product Details