The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005530.2 | NP_005521.1 | 2 - 3 | 115 | 86 |
XM_005254336.2 | XP_005254393.1 | 2 - 3 | 166 | 86 | |
XM_017022119.1 | XP_016877608.1 | 2 - 3 | 335 | 86 | |
GenBank mRNA | AK222973.1 | - | 2 - 3 | 99 | 86 |
AK299970.1 | - | 2 - 3 | 132 | 86 | |
AK315963.1 | - | 2 - 3 | 114 | 86 | |
AK316051.1 | - | 2 - 3 | 102 | 86 | |
BC021967.2 | - | 2 - 3 | 108 | 86 | |
BC036227.1 | - | 2 - 3 | 116 | 86 | |
U07681.1 | - | 2 - 3 | 93 | 86 |