Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
Chromosome Location
Chr.8: 97869021 - 98036720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317748.1 NP_001304677.1 10 - 11 1868 93
NM_002380.4 NP_002371.3 11 - 12 1991 93
NM_030583.3 NP_085072.2 11 - 12 1991 93
XM_005250920.1 XP_005250977.1 11 - 12 1956 93
XM_017013417.1 XP_016868906.1 10 - 11 1833 93
XM_017013418.1 XP_016868907.1 11 - 12 1956 93
GenBank mRNA AK027775.1 - 5 - 6 622 93
AK289721.1 - 11 - 12 2172 93
AL137638.1 - 9 - 10 1053 93
AL833931.1 - 11 - 12 1879 93
AY358895.1 - 10 - 11 1778 93
BC010444.1 - 11 - 12 1920 93
BC016394.1 - 8 - 9 976 93
BX649093.1 - 8 - 9 1045 93
U69263.2 - 10 - 11 1823 93

View Full Product Details