The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002380.4 | NP_002371.3 | 6 - 7 | 1376 | 120 |
NM_030583.3 | NP_085072.2 | 6 - 7 | 1376 | 120 | |
XM_005250920.1 | XP_005250977.1 | 6 - 7 | 1341 | 120 | |
XM_017013418.1 | XP_016868907.1 | 6 - 7 | 1341 | 120 | |
GenBank mRNA | AL137638.1 | - | 4 - 5 | 438 | 120 |
AL833931.1 | - | 6 - 7 | 1264 | 120 | |
BC010444.1 | - | 6 - 7 | 1305 | 120 | |
BC016394.1 | - | 3 - 4 | 361 | 120 | |
BX649093.1 | - | 3 - 4 | 430 | 120 | |
U69263.2 | - | 5 - 6 | 1208 | 120 |