Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
Chromosome Location
Chr.8: 97869021 - 98036720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317748.1 NP_001304677.1 9 - 10 1740 119
NM_002380.4 NP_002371.3 10 - 11 1863 119
NM_030583.3 NP_085072.2 10 - 11 1863 119
XM_005250920.1 XP_005250977.1 10 - 11 1828 119
XM_017013417.1 XP_016868906.1 9 - 10 1705 119
XM_017013418.1 XP_016868907.1 10 - 11 1828 119
GenBank mRNA AK027775.1 - 4 - 5 494 119
AK289721.1 - 10 - 11 2044 119
AL137638.1 - 8 - 9 925 119
AL833931.1 - 10 - 11 1751 119
AY358895.1 - 9 - 10 1650 119
BC010444.1 - 10 - 11 1792 119
BC016394.1 - 7 - 8 848 119
BX649093.1 - 7 - 8 917 119
U69263.2 - 9 - 10 1695 119

View Full Product Details