Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITSN1
Entrez Gene ID
6453
Gene Name
intersectin 1
Gene Aliases
ITSN, SH3D1A, SH3P17
Chromosome Location
Chr.21: 33642400 - 33899861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001132.1 NP_001001132.1 11 - 12 1308 95
NM_003024.2 NP_003015.2 11 - 12 1308 95
XM_005261026.2 XP_005261083.1 11 - 12 1386 95
XM_005261027.1 XP_005261084.1 11 - 12 1386 95
XM_005261031.1 XP_005261088.1 11 - 12 1386 95
XM_011529691.1 XP_011527993.1 11 - 12 1275 95
XM_011529692.1 XP_011527994.1 11 - 12 1275 95
XM_011529693.2 XP_011527995.1 11 - 12 1386 95
XM_017028428.1 XP_016883917.1 12 - 13 1306 95
XM_017028429.1 XP_016883918.1 11 - 12 1275 95
XM_017028430.1 XP_016883919.1 11 - 12 1275 95
XM_017028431.1 XP_016883920.1 11 - 12 1386 95
XM_017028432.1 XP_016883921.1 11 - 12 1386 95
XM_017028433.1 XP_016883922.1 11 - 12 1386 95
XM_017028434.1 XP_016883923.1 11 - 12 1386 95
XM_017028435.1 XP_016883924.1 11 - 12 1275 95
XM_017028436.1 XP_016883925.1 11 - 12 1275 95
XM_017028438.1 XP_016883927.1 11 - 12 1275 95
XM_017028439.1 XP_016883928.1 11 - 12 1386 95
XM_017028440.1 XP_016883929.1 11 - 12 1275 95
XM_017028441.1 XP_016883930.1 11 - 12 1386 95
GenBank mRNA AF114487.1 - 11 - 12 1308 95
AF114488.1 - 11 - 12 1308 95
AK027846.1 - 11 - 12 1275 95
AK074554.1 - 11 - 12 1244 95
AK075290.1 - 11 - 12 1303 95
AK300274.1 - 11 - 12 1283 95
BC013578.1 - 11 - 12 1171 95
BC020269.1 - 11 - 12 1221 95
BC058925.1 - 11 - 12 1335 95
BC073744.1 - 11 - 12 1250 95
BC116185.1 - 11 - 12 1119 95
BC116186.2 - 11 - 12 1119 95
BC117560.1 - 11 - 12 1288 95
BX538175.1 - 6 - 7 607 95
DQ340865.1 - 10 - 11 1036 95
DQ386455.1 - 10 - 11 1059 95
EU107386.1 - 11 - 12 1014 95
EU117382.1 - 10 - 11 1041 95
EU120733.1 - 10 - 11 1041 95
EU120734.1 - 10 - 11 1041 95
EU120735.1 - 11 - 12 1014 95
EU120736.1 - 11 - 12 1014 95
EU140799.1 - 11 - 12 1125 95
EU140800.1 - 10 - 11 930 95
EU152331.1 - 10 - 11 1041 95
EU191901.1 - 10 - 11 964 95
GU939616.1 - 6 - 7 730 95

View Full Product Details