Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITSN1
Entrez Gene ID
6453
Gene Name
intersectin 1
Gene Aliases
ITSN, SH3D1A, SH3P17
Chromosome Location
Chr.21: 33642400 - 33899861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001132.1 NP_001001132.1 21 - 22 2839 66
NM_003024.2 NP_003015.2 21 - 22 2839 66
XM_005261026.2 XP_005261083.1 20 - 21 2902 66
XM_005261027.1 XP_005261084.1 20 - 21 2902 66
XM_005261031.1 XP_005261088.1 20 - 21 2902 66
XM_011529691.1 XP_011527993.1 20 - 21 2791 66
XM_011529692.1 XP_011527994.1 20 - 21 2791 66
XM_011529693.2 XP_011527995.1 20 - 21 2902 66
XM_017028428.1 XP_016883917.1 22 - 23 2837 66
XM_017028429.1 XP_016883918.1 21 - 22 2806 66
XM_017028430.1 XP_016883919.1 20 - 21 2791 66
XM_017028431.1 XP_016883920.1 21 - 22 2917 66
XM_017028432.1 XP_016883921.1 20 - 21 2902 66
XM_017028433.1 XP_016883922.1 21 - 22 2917 66
XM_017028434.1 XP_016883923.1 20 - 21 2902 66
XM_017028435.1 XP_016883924.1 21 - 22 2806 66
XM_017028436.1 XP_016883925.1 20 - 21 2791 66
XM_017028437.1 XP_016883926.1 16 - 17 2257 66
XM_017028438.1 XP_016883927.1 21 - 22 2806 66
XM_017028439.1 XP_016883928.1 21 - 22 2917 66
XM_017028440.1 XP_016883929.1 21 - 22 2806 66
XM_017028441.1 XP_016883930.1 21 - 22 2917 66
GenBank mRNA AF064243.1 - 21 - 22 2677 66
AF064244.1 - 21 - 22 2677 66
AF114487.1 - 21 - 22 2839 66
AF114488.1 - 21 - 22 2839 66
AK300274.1 - 20 - 21 2799 66
AY603419.1 - 2 - 3 274 66
BC116185.1 - 20 - 21 2635 66
BC116186.2 - 21 - 22 2650 66
BC117560.1 - 20 - 21 2804 66
BX538175.1 - 15 - 16 2123 66
DQ340865.1 - 19 - 20 2552 66
DQ679754.1 - 20 - 21 2641 66
DQ679755.1 - 21 - 22 2656 66
DQ679756.1 - 21 - 22 2530 66
DQ679757.1 - 20 - 21 2641 66
EU107386.1 - 21 - 22 2545 66
EU117382.1 - 19 - 20 2557 66
EU120733.1 - 19 - 20 2557 66
EU120734.1 - 19 - 20 2557 66
EU120735.1 - 20 - 21 2530 66
EU120736.1 - 20 - 21 2530 66
EU140799.1 - 20 - 21 2641 66
EU140800.1 - 20 - 21 2461 66
EU152331.1 - 19 - 20 2557 66
EU191901.1 - 19 - 20 2480 66
GU939616.1 - 15 - 16 2246 66
GU939617.1 - 15 - 16 2268 66
U61166.1 - 4 - 5 720 66

View Full Product Details