Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITSN1
Entrez Gene ID
6453
Gene Name
intersectin 1
Gene Aliases
ITSN, SH3D1A, SH3P17
Chromosome Location
Chr.21: 33642400 - 33899861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001132.1 NP_001001132.1 3 - 4 383 124
NM_003024.2 NP_003015.2 3 - 4 383 124
XM_005261026.2 XP_005261083.1 3 - 4 461 124
XM_005261027.1 XP_005261084.1 3 - 4 461 124
XM_005261031.1 XP_005261088.1 3 - 4 461 124
XM_011529691.1 XP_011527993.1 3 - 4 461 124
XM_011529692.1 XP_011527994.1 3 - 4 461 124
XM_011529693.2 XP_011527995.1 3 - 4 461 124
XM_017028428.1 XP_016883917.1 4 - 5 381 124
XM_017028429.1 XP_016883918.1 3 - 4 461 124
XM_017028430.1 XP_016883919.1 3 - 4 461 124
XM_017028431.1 XP_016883920.1 3 - 4 461 124
XM_017028432.1 XP_016883921.1 3 - 4 461 124
XM_017028433.1 XP_016883922.1 3 - 4 461 124
XM_017028434.1 XP_016883923.1 3 - 4 461 124
XM_017028435.1 XP_016883924.1 3 - 4 461 124
XM_017028436.1 XP_016883925.1 3 - 4 461 124
XM_017028438.1 XP_016883927.1 3 - 4 461 124
XM_017028439.1 XP_016883928.1 3 - 4 461 124
XM_017028440.1 XP_016883929.1 3 - 4 461 124
XM_017028441.1 XP_016883930.1 3 - 4 461 124
GenBank mRNA AF064243.1 - 3 - 4 221 124
AF064244.1 - 3 - 4 221 124
AF114487.1 - 3 - 4 383 124
AF114488.1 - 3 - 4 383 124
AK027846.1 - 3 - 4 461 124
AK074554.1 - 3 - 4 319 124
AK075290.1 - 3 - 4 378 124
AK300274.1 - 3 - 4 358 124
BC013578.1 - 3 - 4 246 124
BC020269.1 - 3 - 4 296 124
BC039036.1 - 3 - 4 378 124
BC058925.1 - 3 - 4 410 124
BC073744.1 - 3 - 4 325 124
BC116185.1 - 3 - 4 194 124
BC116186.2 - 3 - 4 194 124
BC117560.1 - 3 - 4 363 124
DQ340865.1 - 2 - 3 111 124
DQ386455.1 - 2 - 3 134 124
DQ679754.1 - 3 - 4 200 124
DQ679756.1 - 3 - 4 200 124
DQ679757.1 - 3 - 4 200 124
EU107386.1 - 3 - 4 200 124
EU117382.1 - 2 - 3 116 124
EU120733.1 - 2 - 3 116 124
EU120734.1 - 2 - 3 116 124
EU120735.1 - 3 - 4 200 124
EU120736.1 - 3 - 4 200 124
EU140799.1 - 3 - 4 200 124
EU140800.1 - 2 - 3 116 124
EU152331.1 - 2 - 3 116 124

View Full Product Details