Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCALD
Entrez Gene ID
83988
Gene Name
neurocalcin delta
Gene Aliases
-
Chromosome Location
Chr.8: 101686542 - 102124907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040624.1 NP_001035714.1 3 - 4 240 131
NM_001040626.1 NP_001035716.1 2 - 3 153 131
XM_017013901.1 XP_016869390.1 2 - 3 343 131
GenBank mRNA AK096515.1 - 3 - 4 165 131
DA185335.1 - 2 - 3 83 131

View Full Product Details