The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040624.1 | NP_001035714.1 | 4 - 5 | 285 | 86 |
NM_001040626.1 | NP_001035716.1 | 3 - 4 | 198 | 86 | |
XM_017013901.1 | XP_016869390.1 | 3 - 4 | 86 | ||
GenBank mRNA | AK096515.1 | - | 4 - 5 | 210 | 86 |
DA185335.1 | - | 3 - 4 | 128 | 86 |