Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC4
Entrez Gene ID
55146
Gene Name
zinc finger DHHC-type containing 4
Gene Aliases
ZNF374
Chromosome Location
Chr.7: 6577413 - 6588996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134387.1 NP_001127859.1 6 - 7 945 69
NM_001134388.1 NP_001127860.1 7 - 8 854 69
NM_001134389.1 NP_001127861.1 6 - 7 723 69
NM_018106.3 NP_060576.1 6 - 7 788 69
XM_005249793.2 XP_005249850.1 7 - 8 898 69
XM_005249794.3 XP_005249851.1 6 - 7 888 69
XM_005249795.2 XP_005249852.1 6 - 7 721 69
XM_005249796.3 XP_005249853.1 6 - 7 835 69
XM_011515442.2 XP_011513744.1 5 - 6 804 69
XM_017012391.1 XP_016867880.1 5 - 6 739 69
GenBank mRNA AF201931.1 - 5 - 6 717 69
AK001341.1 - 6 - 7 704 69
AK223532.1 - 6 - 7 788 69
AL136674.1 - 6 - 7 696 69
AY359090.1 - 6 - 7 876 69
BC001239.1 - 7 - 8 807 69
BQ185277.1 - 2 - 3 69
CR533511.1 - 4 - 5 496 69

View Full Product Details