The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006739.3 | NP_006730.2 | 15 - 16 | 2073 | 62 |
XM_006724242.3 | XP_006724305.1 | 15 - 16 | 2128 | 62 | |
GenBank mRNA | AK122853.1 | - | 14 - 15 | 1921 | 62 |
AK130620.1 | - | 14 - 15 | 2027 | 62 | |
AK223323.1 | - | 15 - 16 | 2063 | 62 | |
AK291136.1 | - | 15 - 16 | 2054 | 62 | |
AK298624.1 | - | 15 - 16 | 1950 | 62 | |
AK312533.1 | - | 15 - 16 | 2053 | 62 | |
BC000142.1 | - | 15 - 16 | 2025 | 62 | |
BC003656.2 | - | 14 - 15 | 1999 | 62 | |
CR456517.1 | - | 15 - 16 | 2013 | 62 | |
D83986.1 | - | 15 - 16 | 2058 | 62 | |
X74795.2 | - | 14 - 15 | 2001 | 62 |