Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCM5
Entrez Gene ID
4174
Gene Name
minichromosome maintenance complex component 5
Gene Aliases
CDC46, P1-CDC46
Chromosome Location
Chr.22: 35400068 - 35454942 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006739.3 NP_006730.2 4 - 5 520 79
XM_006724242.3 XP_006724305.1 4 - 5 575 79
GenBank mRNA AK130620.1 - 3 - 4 474 79
AK223323.1 - 4 - 5 79
AK291136.1 - 4 - 5 501 79
AK298624.1 - 4 - 5 397 79
AK301121.1 - - 662 79
AK312533.1 - 4 - 5 500 79
BC000142.1 - 4 - 5 472 79
BC003656.2 - 3 - 4 446 79
BM787690.1 - 4 - 5 523 79
CR456517.1 - 4 - 5 460 79
D83986.1 - 4 - 5 508 79
X74795.2 - 3 - 4 448 79

View Full Product Details