The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_001204204.2 | NP_001191133.1 | 9 - 10 | 1010 | 87 | |
NM_001291932.1 | NP_001278861.1 | 10 - 11 | 1183 | 87 | ||
NM_012429.4 | NP_036561.1 | 11 - 12 | 1259 | 87 | ||
GenBank mRNA | AB033012.2 | - | 11 - 12 | 1158 | 87 | |
AK056163.1 | - | 5 - 6 | 628 | 87 | ||
AK091402.1 | - | 9 - 10 | 1046 | 87 | ||
AK223587.1 | - | 11 - 12 | 1219 | 87 | ||
AK296540.1 | - | 10 - 11 | 1073 | 87 | ||
AK303751.1 | - | 9 - 10 | 906 | 87 | ||
AK314390.1 | - | 11 - 12 | 1192 | 87 | ||
AL096881.1 | - | 11 - 12 | 1256 | 87 | ||
AY927437.1 | - | LIT3470 AY927437 |
6 - 7 | 593 | 87 | |
CR456571.1 | - | 11 - 12 | 1156 | 87 |