Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO36
Entrez Gene ID
130888
Gene Name
F-box protein 36
Gene Aliases
Fbx36
Chromosome Location
Chr.2: 229922491 - 230013109 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174899.4 NP_777559.3 2 - 3 233 136
XM_005246317.1 XP_005246374.1 2 - 3 243 136
GenBank mRNA AK123085.1 - 3 - 4 538 136
AK289655.1 - 2 - 3 218 136
AK309702.1 - 2 - 3 211 136
BC022538.1 - 3 - 4 349 136
BC033935.1 - 2 - 3 226 136

View Full Product Details