Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
Chromosome Location
Chr.12: 117143473 - 117190500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015002.2 NP_055817.1 11 - 12 1715 76
NM_033624.2 NP_296373.1 11 - 12 1736 76
XM_017019037.1 XP_016874526.1 10 - 11 1561 76
XM_017019038.1 XP_016874527.1 10 - 11 1540 76
XM_017019039.1 XP_016874528.1 10 - 11 1331 76
GenBank mRNA AB020682.1 - 11 - 12 1677 76
AF174601.1 - 11 - 12 1676 76
AK001699.1 - 11 - 12 1699 76
AK074714.1 - 8 - 9 1184 76
AK308141.1 - 12 - 13 1621 76
AK314573.1 - 11 - 12 1688 76
AL136899.1 - 11 - 12 1549 76
BC034045.1 - 12 - 13 1642 76
BC042333.1 - 11 - 12 1435 76
BC091496.1 - 11 - 12 1674 76

View Full Product Details