Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
Chromosome Location
Chr.12: 117143473 - 117190500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015002.2 NP_055817.1 5 - 6 783 72
NM_033624.2 NP_296373.1 5 - 6 783 72
XM_017019037.1 XP_016874526.1 5 - 6 788 72
XM_017019038.1 XP_016874527.1 5 - 6 788 72
XM_017019039.1 XP_016874528.1 4 - 5 399 72
GenBank mRNA AB020682.1 - 5 - 6 745 72
AF174601.1 - 5 - 6 744 72
AK001699.1 - 5 - 6 767 72
AK074714.1 - 2 - 3 252 72
AK308141.1 - 6 - 7 690 72
AK314573.1 - 5 - 6 756 72
AL136899.1 - 5 - 6 596 72
BC034045.1 - 6 - 7 710 72
BC042333.1 - 6 - 7 683 72
BC091496.1 - 5 - 6 742 72

View Full Product Details