Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO21
Entrez Gene ID
23014
Gene Name
F-box protein 21
Gene Aliases
FBX21
Chromosome Location
Chr.12: 117143473 - 117190500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015002.2 NP_055817.1 6 - 7 913 127
NM_033624.2 NP_296373.1 6 - 7 913 127
XM_017019037.1 XP_016874526.1 6 - 7 918 127
XM_017019038.1 XP_016874527.1 6 - 7 918 127
XM_017019039.1 XP_016874528.1 5 - 6 529 127
GenBank mRNA AB020682.1 - 6 - 7 875 127
AF174601.1 - 6 - 7 874 127
AK001699.1 - 6 - 7 897 127
AK074714.1 - 3 - 4 382 127
AK314573.1 - 6 - 7 886 127
AL136899.1 - 6 - 7 726 127
BC034045.1 - 7 - 8 840 127
BC042333.1 - 7 - 8 813 127
BC091496.1 - 6 - 7 872 127

View Full Product Details