Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX54
Entrez Gene ID
79039
Gene Name
DEAD-box helicase 54
Gene Aliases
DP97
Chromosome Location
Chr.12: 113157173 - 113185479 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111322.1 NP_001104792.1 17 - 18 2223 74
NM_024072.3 NP_076977.3 17 - 18 2223 74
GenBank mRNA AF478457.1 - 17 - 18 2223 74
AY148094.1 - 17 - 18 2202 74
BC005848.2 - 17 - 18 2144 74

View Full Product Details