The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111322.1 | NP_001104792.1 | 18 - 19 | 2323 | 85 |
NM_024072.3 | NP_076977.3 | 18 - 19 | 2323 | 85 | |
GenBank mRNA | AY148094.1 | - | 18 - 19 | 2302 | 85 |
BC005848.2 | - | 18 - 19 | 2244 | 85 |