The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111322.1 | NP_001104792.1 | 2 - 3 | 337 | 87 |
NM_024072.3 | NP_076977.3 | 2 - 3 | 337 | 87 | |
GenBank mRNA | AF478457.1 | - | 2 - 3 | 337 | 87 |
AY148094.1 | - | 2 - 3 | 316 | 87 | |
BC005848.2 | - | 2 - 3 | 258 | 87 |