Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEN1
Entrez Gene ID
4221
Gene Name
menin 1
Gene Aliases
MEAI, SCG2
Chromosome Location
Chr.11: 64803514 - 64811294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130804.2 NP_570716.1 1 - 2 262 66
GenBank mRNA AJ297489.1 - 1 - 2 262 66

View Full Product Details