Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC46A3
Entrez Gene ID
283537
Gene Name
solute carrier family 46 member 3
Gene Aliases
FKSG16
Chromosome Location
Chr.13: 28700080 - 28719013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135919.1 NP_001129391.1 6 - 7 1920 147
XM_005266361.2 XP_005266418.1 6 - 7 1598 147

View Full Product Details