Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLT1
Entrez Gene ID
2321
Gene Name
fms related tyrosine kinase 1
Gene Aliases
FLT, FLT-1, VEGFR-1, VEGFR1
Chromosome Location
Chr.13: 28300346 - 28495128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002019.4 NP_002010.2 21 - 22 3244 86
XM_017020485.1 XP_016875974.1 20 - 21 3130 86
GenBank mRNA AB209050.1 - 5 - 6 600 86
AF063657.2 - 21 - 22 2959 86
DQ836394.1 - 7 - 8 784 86
DQ836395.1 - 8 - 9 846 86
EF491868.1 - 3 - 4 493 86
JF509744.1 - 3 - 4 377 86

View Full Product Details