Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLT1
Entrez Gene ID
2321
Gene Name
fms related tyrosine kinase 1
Gene Aliases
FLT, FLT-1, VEGFR-1, VEGFR1
Chromosome Location
Chr.13: 28300346 - 28495128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002019.4 NP_002010.2 26 - 27 3769 97
XM_017020485.1 XP_016875974.1 25 - 26 3655 97
GenBank mRNA AB209050.1 - 10 - 11 1125 97
AF063657.2 - 26 - 27 3484 97
DQ836394.1 - 12 - 13 1309 97
DQ836395.1 - 13 - 14 1371 97
EF491868.1 - 8 - 9 1018 97
EF491869.1 - 5 - 6 805 97
EF491870.1 - 6 - 7 709 97
JF509744.1 - 8 - 9 902 97

View Full Product Details